1-Ethylpyrazole-5-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-Ethylpyrazole-5-boronic Acid Pinacol Ester, 1-Ethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

A271129-10G A271129-1G A271129-250MG A271129-25G A271129-5G
76875-072EA 165.51 USD
76875-072 76875-074 76875-076 76875-078 76875-080
1-Ethylpyrazole-5-boronic acid, pinacol ester 98%
1-Ethylpyrazole-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₂
MW: 222.092 g/mol
MDL Number: MFCD16659009
CAS Number: 1007110-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR