Boc-D-Tyr(2-Br-Z)-OH 98+%

Supplier: Ambeed
Warning

A271063-1G A271063-25G A271063-5G
76875-066EA 10 USD
76875-066 76875-068 76875-070
Boc-D-Tyr(2-Br-Z)-OH 98+%
Boc-D-Tyr(2-Br-Z)-OH

Precaución: for Research Use Only

Formula: C₂₂H₂₄BrNO₇
MDL Number: MFCD00062936
CAS Number: 81189-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR