2-(Ethoxycarbonyl)furan-3-boronic acid 97%

Supplier: Ambeed
Warning

A270691-100MG A270691-1G A270691-250MG A270691-5G
76875-062EA 75 USD
76875-062 76875-064 77883-746 77883-747
2-(Ethoxycarbonyl)furan-3-boronic acid 97%
2-(Ethoxycarbonyl)furan-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₅
MDL Number: MFCD12025973
CAS Number: 1150114-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR