(4-((Furan-2-ylmethoxy)methyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A270635-1G A270635-250MG A270635-5G
76875-056EA 125.19 USD
76875-056 76875-058 76875-060
(4-((Furan-2-ylmethoxy)methyl)phenyl)boronic acid 97%
(4-((Furan-2-ylmethoxy)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃BO₄
MW: 232.04 g/mol
MDL Number: MFCD16198493
CAS Number: 1256358-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR