Ethyl l-thiazolidine-4-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Ethyl thiazolidine-4-carboxylate hydrochloride

A270049-1G A270049-25G A270049-5G
76875-038EA 6.37 USD
76875-038 76875-040 76875-042
Ethyl l-thiazolidine-4-carboxylate hydrochloride 98%
Ethyl l-thiazolidine-4-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂S·ClH
MW: 197.683 g/mol
MDL Number: MFCD09031280
CAS Number: 86028-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR