1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)pyridine
A269584-100MG
A269584-1G
A269584-250MG
A269584-5G
76875-028EA
27.59
USD
76875-028
76875-030
76875-032
76875-034
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 95%
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₀BN₃O₂ MW: 285.16 g/mol |
MDL Number:
MFCD08706029 CAS Number: 864754-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...