tert-Butyl 4-[(4-bromobenzene)sulfonyl]piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-((4-bromophenyl)sulphonyl)piperazine-1-carboxylate

A269037-1G A269037-25G A269037-5G
76875-022EA 21.22 USD
76875-022 76875-024 76875-026
tert-Butyl 4-[(4-bromobenzene)sulfonyl]piperazine-1-carboxylate 98%
tert-Butyl 4-[(4-bromobenzene)sulfonyl]piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₁BrN₂O₄S
MW: 405.307 g/mol
MDL Number: MFCD09746327
CAS Number: 259808-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR