Methyl 2-amino-5-bromo-4-isopropylbenzoate 95%

Supplier: Ambeed
Warning

A268794-1G A268794-250MG A268794-25G A268794-5G
76875-008EA 23 USD
76875-008 76875-010 76875-012 76875-014
Methyl 2-amino-5-bromo-4-isopropylbenzoate 95%
Methyl 2-amino-5-bromo-4-isopropylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₄BrNO₂
MDL Number: MFCD09864815
CAS Number: 1000018-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR