4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Diethylcarbamoyl)-3-fluorophenylboronic acid, (4-(Diethylcarbamoyl)-3-fluorophenyl)boronic acid

A268492-1G A268492-250MG A268492-5G
76875-000EA 120.95 USD
76875-000 76875-002 76875-004
4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid 98%
4-(Diethylcarbamoyl)-3-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₃
MW: 239.051 g/mol
MDL Number: MFCD08436057
CAS Number: 874289-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR