8-Hydroxyquinoline-5-carbaldehyde 95%

Supplier: Ambeed
Warning

A268393-100MG A268393-250MG A268393-1G
76874-990EA 47.75 USD
76874-990 76874-992 77593-662
8-Hydroxyquinoline-5-carbaldehyde 95%
8-Hydroxyquinoline-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.16 g/mol
MDL Number: MFCD03425064
CAS Number: 2598-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR