Phthaloyl Amlodipine 95%

Supplier: Ambeed
Warning

Synonyms: 3-Ethyl 5-Methyl 4-(2-Chlorophenyl)-2-[[2-(1,3-dioxoisoindolin-2-yl)ethoxy]methyl]-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(2-Chlorophenyl)-2-[[2-(1,3-dihydro-1,3-dioxo-2H-isoindol-2-yl)ethoxy]methyl]-1,4-dihydro-6-methyl-3,5-pyridinedicarboxylic acid 3-Ethyl 5-Methyl ester, 3-Ethyl 5-methyl 4-(2-chlorophenyl)-2-((2-(1,3-dioxoisoindolin-2-yl)ethoxy)methyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine besilate impurity A

A267810-25G A267810-5G
76874-972EA 12 USD
76874-972 76874-974
Phthaloyl Amlodipine 95%
Phthaloyl Amlodipine

Precaución: for Research Use Only

Formula: C₂₈H₂₇ClN₂O₇
MDL Number: MFCD07782096
CAS Number: 88150-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR