Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(trifluoromethyl)thiazole-4-carboxylate

A266123-100MG A266123-10G A266123-1G A266123-250MG A266123-5G
76874-960EA 28.65 USD
76874-960 76874-962 76874-964 76874-966 76874-968
Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate 97%
Ethyl-2-(trifluoromethyl)thiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₂S
MW: 225.19 g/mol
MDL Number: MFCD10568298
CAS Number: 133046-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR