(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one 95%

Supplier: Ambeed
Warning

Synonyms: 2,3,4,6-Tetra-O-(trimethylsilyl)-D-glucono-delta-lactone, 2,3,4,6-tetra-O-(trimethylsilyl)-D-glucono-1,5-lactone, 2,3,4,6-tetrakis-O-trimethylsilyl-D-gluconolactone

A265675-25G A265675-500G A265675-5G A265675-100G A265675-10G
76874-946EA 13 USD
76874-946 76874-948 76874-950 77568-522 77672-376
(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one 95%
(3R,4S,5R,6R)-3,4,5-tris(trimethylsilyl)oxy)-6-((trimethylsilyl)oxy)methyl)tetrahydro-2H-pyran-2-one

Precaución: For research use only.

Formula: C₁₈H₄₂O₆Si₄
MDL Number: MFCD22665922
CAS Number: 32384-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR