2-Amino-5-methylbenzenesulfonic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Aminotoluene-3-sulphonic acid
A263162-25G
A263162-500G
76874-926EA
5.31
USD
76874-926
76874-928
2-Amino-5-methylbenzenesulfonic acid 98%
2-Amino-5-methylbenzenesulphonic acid
Precaución: for Research Use Only
Formula:
C₇H₉NO₃S MW: 187.22 g/mol |
MDL Number:
MFCD00007908 CAS Number: 88-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...