4-Chloro-3-hydroxyphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-3-hydroxyphenyl)boronic acid

A263017-100MG A263017-1G A263017-250MG
76874-916EA 75.33 USD
76874-916 76874-918 76874-920
4-Chloro-3-hydroxyphenylboronic acid 95%
4-Chloro-3-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₆BClO₃
MW: 172.3740 g/mol
MDL Number: MFCD10697413
CAS Number: 915201-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR