6-Chloro-5-fluoronicotinic acid 95%

Supplier: Ambeed
Warning

A262949-100MG A262949-1G A262949-250MG A262949-5G A262949-25G
76874-910EA 16 USD
76874-910 76874-912 76874-914 77593-634 77883-728
6-Chloro-5-fluoronicotinic acid 95%
6-Chloro-5-fluoronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃ClFNO₂
MDL Number: MFCD11226507
CAS Number: 38186-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR