1-Boc-2-phenyl-4-piperidinone 97%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-2-Phenyl-4-piperidinone

A262568-100MG A262568-10G A262568-1G A262568-250MG
76874-902EA 23.34 USD
76874-902 76874-904 76874-906 76874-908
1-Boc-2-phenyl-4-piperidinone 97%
1-Boc-2-phenyl-4-piperidinone

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₃
MW: 275.3400 g/mol
MDL Number: MFCD04035609
CAS Number: 849928-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR