(2S)-2-((tert-Butoxycarbonyl)amino)-2-(3-hydroxyadamantan-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: Saxagliptin Impurity 23

A262384-10G A262384-1G A262384-250MG A262384-25G A262384-5G
76874-892EA 70.02 USD
76874-892 76874-894 76874-896 76874-898 76874-900
(2S)-2-((tert-Butoxycarbonyl)amino)-2-(3-hydroxyadamantan-1-yl)acetic acid 97%
(2S)-2-((tert-Butoxycarbonyl)amino)-2-(3-hydroxyadamantan-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₇NO₅
MW: 325.4 g/mol
MDL Number: MFCD16660445
CAS Number: 361442-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR