5-Phenylthiophen-2-amine 95%

Supplier: Ambeed
Warning

A261937-100MG A261937-250MG A261937-1G
76874-884EA 118.83 USD
76874-884 76874-886 77672-336
5-Phenylthiophen-2-amine 95%
5-Phenylthiophen-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉NS
MW: 175.25 g/mol
MDL Number: MFCD11904083
CAS Number: 14770-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR