Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-3-methoxy-3,3-diphenylpropanoate

A261861-100MG A261861-1G A261861-250MG A261861-5G
76874-876EA 6 USD
76874-876 76874-878 76874-880 76874-882
Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate 97%
Methyl-2-hydroxy-3-methoxy-3,3-diphenylpropanoate

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₄
MW: 286.33 g/mol
MDL Number: MFCD09833706
CAS Number: 178306-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR