4-Bromo-N,N-diethyl-3-methylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A261557-1G A261557-250MG A261557-5G
76874-868EA 122.01 USD
76874-868 76874-870 76874-872
4-Bromo-N,N-diethyl-3-methylbenzenesulfonamide 98%
4-Bromo-N,N-diethyl-3-methylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrNO₂S
MW: 306.21 g/mol
MDL Number: MFCD12650342
CAS Number: 1178100-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR