4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%

Supplier: Ambeed
Warning

A261074-10G A261074-1G A261074-250MG A261074-5G
76874-860EA 327 USD
76874-860 76874-862 76874-864 76874-866
4-Bromo-N,N-dimethyl-2-fluorobenzamide 98%
4-Bromo-N,N-dimethyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO
MDL Number: MFCD09878360
CAS Number: 749927-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR