4-n-Pentylphenyl-4-pentylbenzoate 97%

Supplier: Ambeed
Warning

A260886-100G A260886-10G A260886-25G A260886-5G A260886-1G
76874-852EA 203.7 USD
76874-852 76874-854 76874-856 76874-858 77612-494
4-n-Pentylphenyl-4-pentylbenzoate 97%
4-n-Pentylphenyl-4-pentylbenzoate

Precaución: for Research Use Only

Formula: C₂₃H₃₀O₂
MW: 338.48 g/mol
MDL Number: MFCD01076318
CAS Number: 74305-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR