2-Amino-4-chlorophenylboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A259970-100MG A259970-250MG A259970-5G A259970-1G
76874-838EA 21.22 USD
76874-838 76874-840 76874-842 77571-600
2-Amino-4-chlorophenylboronic acid, pinacol ester 97%
2-Amino-4-chlorophenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BClNO₂
MW: 253.533 g/mol
MDL Number: MFCD11617915
CAS Number: 863578-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR