5-(2-Bromoacetyl)-2-hydroxybenzaldehyde 97%

Supplier: Ambeed
Danger

A259286-100G A259286-10G A259286-1G A259286-250MG A259286-25G A259286-5G
76874-812EA 335.25 USD
76874-812 76874-814 76874-816 76874-818 76874-820 76874-822
5-(2-Bromoacetyl)-2-hydroxybenzaldehyde 97%
5-(2-Bromoacetyl)-2-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrO₃
MW: 243.05 g/mol
MDL Number: MFCD09833486
CAS Number: 115787-50-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR