(S)-1-tert-Butyl 2-methyl 4-methylenepyrrolidine-1,2-dicarboxylate 97%

Supplier: Ambeed
Warning

A259274-1G A259274-250MG A259274-5G A259274-25G
76874-808EA 85.94 USD
76874-808 76874-810 77568-498 77672-316
(S)-1-tert-Butyl 2-methyl 4-methylenepyrrolidine-1,2-dicarboxylate 97%
(S)-1-tert-Butyl 2-methyl 4-methylenepyrrolidine-1,2-dicarboxylate

Precaución: For research use only.

Formula: C₁₂H₁₉NO₄
MW: 241.28 g/mol
MDL Number: MFCD15832853
CAS Number: 84348-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR