(5-Bromo-2-fluorophenyl)methanamine 98%

Supplier: Ambeed
Warning

A259245-1G A259245-5G A259245-250MG A259245-25G
76874-806EA 30 USD
76874-806 77672-312 77883-710 77883-711
(5-Bromo-2-fluorophenyl)methanamine 98%
(5-Bromo-2-fluorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD06212854
CAS Number: 190656-34-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR