5-(Methoxycarbonyl)furan-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A259026-100MG A259026-50MG A259026-250MG
76874-802EA 202 USD
76874-802 76874-804 77612-478
5-(Methoxycarbonyl)furan-2-carboxylic acid 95%
5-(Methoxycarbonyl)furan-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₅
MDL Number: MFCD19441220
CAS Number: 6750-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR