S-Methylisothiouronium sulfate 97%

Supplier: Ambeed
Warning

A258497-10G A258497-25G A258497-500G A258497-1000G
76874-792EA 5 USD
76874-792 76874-794 76874-796 77672-304
S-Methylisothiouronium sulfate 97%
S-Methylisothiouronium sulphate

Precaución: for Research Use Only

Formula: C₄H₁₂N₄S₂·H₂SO₄
MDL Number: MFCD00013055
CAS Number: 867-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR