Methyl 1-methyl-1H-benzo[d]Imidazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A257917-1G A257917-5G
76874-772EA 108.22 USD
76874-772 76874-776
Methyl 1-methyl-1H-benzo[d]Imidazole-6-carboxylate 97%
Methyl 1-methyl-1H-benzo[d]Imidazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₂
MW: 190.19 g/mol
MDL Number: MFCD11973677
CAS Number: 131020-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR