1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 97%

Supplier: Ambeed
Warning

A257497-100G A257497-10G A257497-1G A257497-25G A257497-5G
76874-752EA 1094.85 USD
76874-752 76874-754 76874-756 76874-758 76874-760
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene 97%
1-Bromo-2-chloro-5-fluoro-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrClFNO₂
MW: 254.4400 g/mol
MDL Number: MFCD11110551
CAS Number: 1027833-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR