(±)-β-Aminophenethyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

A257380-100G A257380-10G A257380-1G A257380-25G A257380-500G A257380-5G
76874-740EA 67.9 USD
76874-740 76874-742 76874-744 76874-746 76874-748 76874-750
(±)-β-Aminophenethyl alcohol 98%
(±)-β-Aminophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00130145
CAS Number: 7568-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR