(S)-2-Methylpiperidine hydrochloride 95%

Supplier: Ambeed
Warning

A256932-100MG A256932-250MG
76874-730EA 17 USD
76874-730 76874-732
(S)-2-Methylpiperidine hydrochloride 95%
(S)-2-Methylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃N·ClH
MDL Number: MFCD26125912
CAS Number: 205526-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR