2-(3,5-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A256535-100MG A256535-1G A256535-250MG A256535-5G
76874-720EA 33 USD
76874-720 76874-722 76874-724 76874-726
2-(3,5-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(3,5-Dichloro-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BCl₂O₂
MDL Number: MFCD12407205
CAS Number: 942069-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR