Benzyl (S)-(-)-1,2,3,4-tetrahydro-3-isoquinolinecarboxylate p-toluenesulfonic acid salt 98%

Supplier: Ambeed
Danger

Synonyms: Benzyl (S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinecarboxylate p-Toluenesulphonic acid salt

A255096-10G A255096-1G A255096-5G
76874-712EA 9.55 USD
76874-712 76874-714 76874-716
Benzyl (S)-(-)-1,2,3,4-tetrahydro-3-isoquinolinecarboxylate p-toluenesulfonic acid salt 98%
Benzyl (S)-(-)-1,2,3,4-tetrahydro-3-isoquinolinecarboxylate p-toluenesulfonic acid salt

Precaución: for Research Use Only

Formula: C₂₄H₂₅NO₅S
MW: 439.5240 g/mol
MDL Number: MFCD00799478
CAS Number: 77497-97-3
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR