2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 95%
Supplier: Ambeed
|
Warning
|
A255091-100MG
A255091-1G
A255091-250MG
A255091-5G
76874-704EA
55.17
USD
76874-704
76874-706
76874-708
76874-710
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid) 95%
2,2'-(Ethane-1,2-diylbis(azanediyl))bis(2-(2-hydroxyphenyl)acetic acid)
Precaución: for Research Use Only
Formula:
C₁₈H₂₀N₂O₆ MW: 360.36 g/mol |
MDL Number:
MFCD00004247 CAS Number: 1170-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...