5-Bromoquinazoline-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A255032-1G A255032-250MG
76874-700EA 440.28 USD
76874-700 76874-702
5-Bromoquinazoline-2,4(1H,3H)-dione 97%
5-Bromoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
MDL Number: MFCD09954847
CAS Number: 959237-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR