6-Bromo-3-iodoimidazo[1,2-a]pyridine 96%

Supplier: Ambeed
Warning

A254990-1G A254990-250MG A254990-5G
76874-694EA 147.47 USD
76874-694 76874-696 76874-698
6-Bromo-3-iodoimidazo[1,2-a]pyridine 96%
6-Bromo-3-iodoimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MW: 322.9300 g/mol
MDL Number: MFCD18800553
CAS Number: 474706-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR