tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutyl)carbamate 98%

Supplier: Ambeed
Warning

A254245-100MG A254245-1G A254245-250MG A254245-5G
76874-680EA 35 USD
76874-680 76874-682 76874-684 77883-689
tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutyl)carbamate 98%
tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₃₂BNO₄
MDL Number: MFCD18762017
CAS Number: 1032528-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR