3-Amino-6-(aminomethyl)-1,2,4-triazin-5(4H)-one acetate 97%

Supplier: Ambeed
Warning

A253641-1G A253641-250MG
76874-676EA 55.17 USD
76874-676 76874-678
3-Amino-6-(aminomethyl)-1,2,4-triazin-5(4H)-one acetate 97%
3-Amino-6-(aminomethyl)-1,2,4-triazin-5(4H)-one acetate

Precaución: for Research Use Only

Formula: C₆H₁₁N₅O₃
MW: 201.18 g/mol
MDL Number: MFCD19703225
CAS Number: 1353100-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR