Methyl 3-(2,4-dihydroxyphenyl)propionate 95%

Supplier: Ambeed
Warning

A252229-1MG A252229-1G A252229-250MG A252229-5G
76874-652EA 6 USD
76874-652 77672-256 77672-258 77883-686
Methyl 3-(2,4-dihydroxyphenyl)propionate 95%
Methyl 3-(2,4-dihydroxyphenyl)propionate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MDL Number: MFCD19441070
CAS Number: 17422-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR