Myoseverin 98%

Supplier: Ambeed
Warning

Synonyms: 9-Isopropyl-2,6-bis(4-methoxybenzylamino)purine, 9-Isopropyl-N2,N6-bis(4-methoxybenzyl)-9H-purine-2,6-diamine

A251062-10MG A251062-1MG A251062-25MG A251062-5MG
76874-624EA 62.6 USD
76874-624 76874-626 76874-628 76874-630
Myoseverin 98%
Myoseverin

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₆O₂
MW: 432.53 g/mol
MDL Number: MFCD02683596
CAS Number: 267402-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR