Boc-4-amino-3-hydroxybutanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

A250490-250MG A250490-1G
76874-618EA 53 USD
76874-618 77883-675
Boc-4-amino-3-hydroxybutanoic acid 95%
Boc-4-amino-3-hydroxybutanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MDL Number: MFCD02682574
CAS Number: 69489-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR