4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Cyclohexyl(methyl)carbamoyl)phenyl)boronic acid

A250259-1G A250259-250MG A250259-5G
76874-612EA 110.34 USD
76874-612 76874-614 76874-616
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid 98%
4-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MW: 261.1300 g/mol
MDL Number: MFCD09027224
CAS Number: 913835-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR