4-Chlor-3-(propylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-3-(propylcarbamoyl)phenyl)boronic acid

A249645-1G A249645-250MG A249645-5G
76874-596EA 162 USD
76874-596 76874-598 76874-600
4-Chlor-3-(propylcarbamoyl)phenylboronic acid 98%
4-Chlor-3-(propylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃BClNO₃
MDL Number: MFCD07783885
CAS Number: 871332-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR