2-Methyl-5-(morpholin-4-ylsulfonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(3-Borono-4-methylbenzenesulphonyl)morpholine, (2-Methyl-5-(morpholinosulphonyl)phenyl)boronic acid

A249322-100MG A249322-1G A249322-250MG A249322-5G
76874-588EA 37 USD
76874-588 76874-590 76874-592 76874-594
2-Methyl-5-(morpholin-4-ylsulfonyl)benzeneboronic acid 98%
2-Methyl-5-(morpholin-4-ylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₅S
MW: 285.124 g/mol
MDL Number: MFCD07363761
CAS Number: 871329-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR