6-Cyano-1H-indole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A249313-100MG A249313-1G A249313-250MG A249313-5G
76874-580EA 16 USD
76874-580 76874-582 76874-584 76874-586
6-Cyano-1H-indole-3-carboxylic acid 98%
6-Cyano-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₂
MDL Number: MFCD09954938
CAS Number: 174500-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR