2,4-Dimethoxy-5,6-dimethylpyrimidine 95%

Supplier: Ambeed
Warning

A249253-100MG A249253-250MG
76874-576EA 10 USD
76874-576 76874-578
2,4-Dimethoxy-5,6-dimethylpyrimidine 95%
2,4-Dimethoxy-5,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD22987595
CAS Number: 120129-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR