Methyl 4-amino-5-iodo-2-methylbenzoate 97%

Supplier: Ambeed
Warning

A248886-250MG A248886-100MG A248886-1G
76874-568EA 35 USD
76874-568 77885-652 77885-653
Methyl 4-amino-5-iodo-2-methylbenzoate 97%
Methyl 4-amino-5-iodo-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀INO₂
MDL Number: MFCD11112906
CAS Number: 672293-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR