Ethyl-8-bromo-4-chloroquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 8-bromo-4-chloroquinoline-3-carboxylate, 8-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

A248771-1G A248771-250MG
76874-562EA 71.09 USD
76874-562 76874-564
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate 97%
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₉BrClNO₂
MW: 314.563 g/mol
MDL Number: MFCD00173385
CAS Number: 206258-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR